Assay Details
Gene Symbol: FAM50A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034496.2 | NP_001029668.2 | 1 - 2 | 182 | 78 | |
GenBank mRNA | BC103028.1 | - | 1 - 2 | 183 | 78 | |
DV888640.1 | - | 1 - 2 | 173 | 78 | ||
EE364565.1 | - | 1 - 2 | 190 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
