Assay Details
Gene Symbol: MRPL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035029.2 | NP_001030201.2 | 2 - 3 | 330 | 96 | |
GenBank mRNA | BC110017.1 | - | 2 - 3 | 330 | 96 | |
BT021548.1 | - | 2 - 3 | 314 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
