Assay Details
Gene Symbol: HMOX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035087.2 | NP_001030259.2 | 5 - 6 | 1015 | 70 | |
GenBank mRNA | DV892281.1 | - | 4 - 5 | 905 | 70 | |
DV912958.1 | - | 2 - 3 | 211 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
