Assay Details
Gene Symbol: MYC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046074.2 | NP_001039539.1 | 2 - 3 | 1161 | 106 | |
GenBank mRNA | BC113343.1 | - | 2 - 3 | 1140 | 106 | |
DV798027.1 | - | 1 - 2 | 484 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
