Assay Details
Gene Symbol: SELENOS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046114.3 | NP_001039579.2 | 1 - 2 | 170 | 63 | |
GenBank mRNA | BC112740.1 | - | 1 - 2 | 152 | 63 | |
DV822816.1 | - | 1 - 2 | 171 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
