Assay Details
Gene Symbol: GSTO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075214.2 | NP_001068682.2 | 1 - 2 | 117 | 87 | |
GenBank mRNA | BC120211.1 | - | 1 - 2 | 103 | 87 | |
EE226879.1 | - | 1 - 2 | 117 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
