Assay Details
Gene Symbol: GSTO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075214.2 | NP_001068682.2 | 2 - 3 | 225 | 126 | |
GenBank mRNA | BC120211.1 | - | 2 - 3 | 211 | 126 | |
EE226879.1 | - | 2 - 3 | 225 | 126 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
