Assay Details
Gene Symbol: GSTO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075214.2 | NP_001068682.2 | 3 - 4 | 440 | 98 | |
GenBank mRNA | BC120211.1 | - | 3 - 4 | 426 | 98 | |
DV852688.1 | - | 1 - 2 | 175 | 98 | ||
EE226879.1 | - | 3 - 4 | 440 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
