Assay Details
Gene Symbol: TARBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075678.2 | NP_001069146.2 | 1 - 2 | 169 | 97 | |
GenBank mRNA | BC122653.1 | - | 1 - 2 | 169 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
