Assay Details
Gene Symbol: TARBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075678.2 | NP_001069146.2 | 5 - 6 | 604 | 79 | |
GenBank mRNA | BC122653.1 | - | 5 - 6 | 604 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
