Assay Details
Gene Symbol: NRAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097989.1 | NP_001091458.1 | 1 - 2 | 107 | 83 | |
GenBank mRNA | BC140659.1 | - | 1 - 2 | 107 | 83 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC140659.1, NM_001097989.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
