Assay Details
Gene Symbol: NRAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097989.1 | NP_001091458.1 | 2 - 3 | 230 | 98 | |
GenBank mRNA | BC140659.1 | - | 2 - 3 | 230 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
