Assay Details
Gene Symbol: EHD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098004.1 | NP_001091473.1 | 1 - 2 | 371 | 60 | |
GenBank mRNA | BC140522.1 | - | 1 - 2 | 371 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
