Assay Details
Gene Symbol: EHD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098004.1 | NP_001091473.1 | 2 - 3 | 540 | 70 | |
GenBank mRNA | BC140522.1 | - | 2 - 3 | 540 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
