Assay Details
Gene Symbol: SMIM19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098040.2 | NP_001091509.2 | 3 - 4 | 402 | 101 | |
NM_001146118.1 | NP_001139590.1 | 3 - 4 | 328 | 101 | ||
GenBank mRNA | BC140495.1 | - | 3 - 4 | 300 | 101 | |
DV927834.1 | - | 3 - 4 | 402 | 101 | ||
EE349132.1 | - | 3 - 4 | 393 | 101 | ||
EE386282.1 | - | 3 - 4 | 285 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
