Assay Details
Gene Symbol: RPS7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098874.2 | NP_001092344.1 | 3 - 4 | 211 | 113 | |
GenBank mRNA | BC146133.1 | - | 3 - 4 | 213 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
