Assay Details
Gene Symbol: BLOC1S5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098875.1 | NP_001092345.1 | 4 - 5 | 399 | 74 | |
GenBank mRNA | BC146091.1 | - | 4 - 5 | 399 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
