Assay Details
Gene Symbol: DUSP19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098878.1 | NP_001092348.1 | 2 - 3 | 410 | 148 | |
GenBank mRNA | BC146174.1 | - | 2 - 3 | 410 | 148 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
