Assay Details
Gene Symbol: CILK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098887.1 | NP_001092357.1 | 11 - 12 | 1878 | 72 | |
GenBank mRNA | BC142307.1 | - | 11 - 12 | 1878 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
