Assay Details
Gene Symbol: CILK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098887.1 | NP_001092357.1 | 12 - 13 | 2008 | 84 | |
GenBank mRNA | BC142307.1 | - | 12 - 13 | 2008 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
