Assay Details
Gene Symbol: DTYMK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098893.1 | NP_001092363.1 | 4 - 5 | 549 | 84 | |
GenBank mRNA | BC142258.1 | - | 4 - 5 | 549 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
