Assay Details
Gene Symbol: RNF135 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098936.1 | NP_001092406.1 | 1 - 2 | 367 | 153 | |
GenBank mRNA | BC146080.1 | - | 1 - 2 | 367 | 153 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
