Assay Details
Gene Symbol: RNF135 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098936.1 | NP_001092406.1 | 3 - 4 | 670 | 82 | |
GenBank mRNA | BC146080.1 | - | 3 - 4 | 670 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
