Assay Details
Gene Symbol: MLYCD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098946.1 | NP_001092416.1 | 2 - 3 | 671 | 79 | |
GenBank mRNA | BC142049.1 | - | 2 - 3 | 671 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
