Assay Details
Gene Symbol: CDKN1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098958.2 | NP_001092428.1 | 1 - 2 | 98 | 75 | |
GenBank mRNA | BC142538.1 | - | 1 - 2 | 99 | 75 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC142538.1, NM_001098958.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
