Assay Details
Gene Symbol: TAF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098968.2 | NP_001092438.1 | 2 - 3 | 413 | 72 | |
GenBank mRNA | BC142260.1 | - | 2 - 3 | 414 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
