Assay Details
Gene Symbol: MLEC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098983.1 | NP_001092453.1 | 3 - 4 | 709 | 79 | |
GenBank mRNA | BC110237.1 | - | 3 - 4 | 704 | 79 | |
BC146164.1 | - | 3 - 4 | 709 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
