Assay Details
Gene Symbol: MTREX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099012.1 | NP_001092482.1 | 23 - 24 | 2771 | 75 | |
GenBank mRNA | BC142299.1 | - | 23 - 24 | 2771 | 75 | |
BC146077.1 | - | 23 - 24 | 2922 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
