Assay Details
Gene Symbol: PINK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099701.2 | NP_001093171.1 | 3 - 4 | 810 | 146 | |
GenBank mRNA | BC142192.1 | - | 3 - 4 | 811 | 146 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
