Assay Details
Gene Symbol: DYRK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100298.1 | NP_001093768.1 | 1 - 2 | 188 | 112 | |
GenBank mRNA | BC149927.1 | - | 1 - 2 | 188 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
