Assay Details
Gene Symbol: FKBP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100330.2 | NP_001093800.1 | 3 - 4 | 574 | 84 | |
GenBank mRNA | BC149352.1 | - | 3 - 4 | 576 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
