Assay Details
Gene Symbol: PRADC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100338.2 | NP_001093808.1 | 4 - 5 | 521 | 74 | |
GenBank mRNA | BC150018.1 | - | 4 - 5 | 522 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
