Assay Details
Gene Symbol: SRP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100350.1 | NP_001093820.1 | 1 - 2 | 91 | 61 | |
GenBank mRNA | BC149893.1 | - | 1 - 2 | 91 | 61 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC149893.1, NM_001100350.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
