Assay Details
Gene Symbol: SRP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100350.1 | NP_001093820.1 | 4 - 5 | 307 | 71 | |
GenBank mRNA | BC149893.1 | - | 4 - 5 | 307 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
