Assay Details
Gene Symbol: C1QTNF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101872.2 | NP_001095342.1 | 1 - 2 | 120 | 110 | |
GenBank mRNA | BC151625.1 | - | 1 - 2 | 121 | 110 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC151625.1, NM_001101872.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
