Assay Details
Gene Symbol: SHOC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101943.1 | NP_001095413.1 | 6 - 7 | 1538 | 142 | |
GenBank mRNA | BC148097.1 | - | 6 - 7 | 1538 | 142 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
