Assay Details
Gene Symbol: SHOC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101943.1 | NP_001095413.1 | 8 - 9 | 1800 | 111 | |
GenBank mRNA | BC148097.1 | - | 8 - 9 | 1800 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
