Assay Details
Gene Symbol: MAP3K11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101991.1 | NP_001095461.1 | 1 - 2 | 1202 | 64 | |
GenBank mRNA | BC149999.1 | - | 1 - 2 | 1202 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
