Assay Details
Gene Symbol: RHBDL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102014.2 | NP_001095484.1 | 5 - 6 | 731 | 71 | |
GenBank mRNA | BC149360.1 | - | 5 - 6 | 732 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
