Assay Details
Gene Symbol: ADH4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102059.2 | NP_001095529.1 | 1 - 2 | 101 | 71 | |
GenBank mRNA | BC149290.1 | - | 1 - 2 | 102 | 71 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC149290.1, NM_001102059.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
