Assay Details
Gene Symbol: TIPIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105336.3 | NP_001098806.2 | 2 - 3 | 145 | 86 | |
GenBank mRNA | BC102519.1 | - | 2 - 3 | 146 | 86 | |
EH146865.1 | - | 2 - 3 | 146 | 86 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC102519.1, NM_001105336.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
