Assay Details
Gene Symbol: HDGFL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105361.1 | NP_001098831.1 | 5 - 6 | 651 | 68 | |
GenBank mRNA | BC153286.1 | - | 5 - 6 | 651 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
