Assay Details
Gene Symbol: COMTD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105368.2 | NP_001098838.1 | 5 - 6 | 619 | 92 | |
GenBank mRNA | BC134766.1 | - | 5 - 6 | 620 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
