Assay Details
Gene Symbol: BAG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110532.2 | NP_001104002.1 | 3 - 4 | 682 | 59 | |
GenBank mRNA | BT025395.1 | - | 3 - 4 | 626 | 59 | |
BT030482.1 | - | 3 - 4 | 626 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
