Assay Details
Gene Symbol: KANSL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024571.2 | NP_001019742.2 | 8 - 9 | 1165 | 68 | |
GenBank mRNA | BC111221.1 | - | 8 - 9 | 1165 | 68 | |
BT021786.1 | - | 8 - 9 | 1199 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
