Assay Details
Gene Symbol: C1QTNF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076201.3 | NP_001069669.3 | 1 - 2 | 220 | 82 | |
GenBank mRNA | BC120196.1 | - | 1 - 2 | 220 | 82 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC120196.1, NM_001076201.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
