Assay Details
Gene Symbol: ARL2BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076274.2 | NP_001069742.2 | 3 - 4 | 495 | 101 | |
GenBank mRNA | BC108166.1 | - | 3 - 4 | 311 | 101 | |
BC134728.1 | - | 3 - 4 | 495 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
