Assay Details
Gene Symbol: USP12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098059.1 | NP_001091528.1 | 5 - 6 | 715 | 120 | |
GenBank mRNA | BT030596.2 | - | 5 - 6 | 713 | 120 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
