Assay Details
Gene Symbol: CASC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098069.1 | NP_001091538.1 | 13 - 14 | 2190 | 114 | |
GenBank mRNA | BC140556.1 | - | 13 - 14 | 2190 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
