Assay Details
Gene Symbol: YARS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098088.2 | NP_001091557.1 | 1 - 2 | 877 | 105 | |
GenBank mRNA | BC140541.1 | - | 1 - 2 | 878 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
